Search
Search

Wdr48| Gene Symbol: Wdr48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026236.3 | NP_080512.1 | 9, 10 | 996 | ||
| GenBank mRNA | AK012599.1 | - | 9, 10 | 996 | ||
| AK018396.1 | - | 9, 10 | 999 | |||
| AK030041.1 | - | 9, 10 | 997 | |||
| AK031244.1 | - | 9, 10 | 1009 | |||
| AK033631.1 | - | 9, 10 | 995 | |||
| AK035273.1 | - | 9, 10 | 998 | |||
| AK035864.1 | - | 9, 10 | 998 | |||
| AK043970.1 | - | 9, 10 | 998 | |||
| AK122511.1 | - | 9, 10 | 980 | |||
| AK128953.1 | - | 9, 10 | 996 | |||
| BC006679.1 | - | 8, 9 | 859 | |||
| BC040795.1 | - | 5, 6 | 527 | |||
| BC048155.1 | - | 9, 10 | 985 | 5698339 | ||
| BC050106.1 | - | 9, 10 | 1013 | |||
| BC062967.1 | - | 9, 10 | 1000 | 6835444 | ||