Search
Search

Rai14| Gene Symbol: Rai14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166408.1 | NP_001159880.1 | 14 | 1232 | ||
| NM_030690.3 | NP_109615.1 | 14 | 1318 | |||
| GenBank mRNA | AF202315.1 | - | 13 | 1105 | ||
| AF274866.1 | - | 14 | 1297 | |||
| AK129333.1 | - | 14 | 1168 | |||
| AK141233.1 | - | 10 | 818 | |||
| AK160119.1 | - | 13 | 1141 | |||
| BC052458.1 | - | 14 | 1318 | 6830677 | ||