Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023131.1 | NP_075620.1 | 6 | 639 | ||
GenBank mRNA | AJ251645.1 | - | 6 | 639 | ||
AK012946.1 | - | 6 | 595 | |||
BC004060.1 | - | 5 | 488 | 3590170 | ||
BC048686.1 | - | 6 | 688 |