Search
Search

Zbtb17| Gene Symbol: Zbtb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009541.2 | NP_033567.2 | 16 | 2374 | ||
| GenBank mRNA | AK051971.1 | - | 15 | 2445 | ||
| AK134251.1 | - | 16 | 2334 | |||
| BC060055.1 | - | 13 | 2644 | |||
| BC145312.1 | - | 15 | 2247 | 9053867 | ||
| BC145313.1 | - | 13 | 2453 | |||
| BC145821.1 | - | 15 | 2247 | 40131124 | ||
| U14556.1 | - | 16 | 2372 | |||
| U22396.1 | - | 16 | 2362 | |||