Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nelfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045863.1 | NP_001039328.1 | 10 | 1213 | ||
NM_001045864.1 | NP_001039329.1 | 10 | 1299 | |||
NM_138580.2 | NP_613046.2 | 10 | 1087 | |||
GenBank mRNA | AI325150.1 | - | 2 | 319 | ||
AK011663.1 | - | 10 | 1072 | |||
AK012389.1 | - | 10 | 1104 | |||
AK131947.1 | - | 10 | 1071 | |||
AK147011.1 | - | 10 | 1195 | |||
BC034867.1 | - | 10 | 1039 | 1495547 |