Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rspo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138683.2 | NP_619624.2 | 3 | 620 | ||
GenBank mRNA | AB016768.1 | - | 3 | 620 | ||
AK132277.1 | - | 3 | 567 | |||
BC138831.1 | - | 2 | 372 | 8861853 | ||
BC138833.1 | - | 2 | 372 | 8861855 |