Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cbln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019820.3 | NP_062794.1 | 1, 2 | 342 | ||
GenBank mRNA | AF218379.1 | - | 1, 2 | 318 | ||
AK081085.1 | - | 1, 2 | 317 | |||
BC065108.1 | - | 1, 2 | 342 | 30536445 |