Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027544.2 | NP_081820.2 | 1 | 550 | ||
GenBank mRNA | AB250379.1 | - | 1 | 433 | ||
AK077129.1 | - | 1 | 433 | |||
AY141192.1 | - | 1 | 550 | |||
BC049764.1 | - | 1 | 411 | 6774330 |