Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wnt9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139298.2 | NP_647459.1 | 2, 3 | 400 | ||
GenBank mRNA | AB072311.1 | - | 2, 3 | 364 | ||
AK087490.1 | - | 2, 3 | 403 | |||
BC066165.1 | - | 2, 3 | 389 | 30435371 |