Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOX3 SIRNA Assays ›
Gene Symbol:
Nox3
Gene Name:
NADPH oxidase 3
Gene Aliases:
GP91-3, het, nmf250
Chromosome Location:
Chr.17: 3635240 - 3696261 on Build GRCm38
UniGene ID:
Mm.328756
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS278876
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nox3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_198958.2
NP_945196.2
6
689
GenBank mRNA
AY182377.1
-
6
689
AY573240.1
-
6
629
BC106862.2
-
6
689
40047310
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
detection of gravity
response to gravity
superoxide anion generation
otolith development
oxidation-reduction process
protein binding
superoxide-generating NADPH oxidase activity
oxidoreductase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bfkrn:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0