Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: H2bfm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027067.2 | NP_081343.1 | 1 | 596 | ||
GenBank mRNA | AK005982.1 | - | 1 | 597 | ||
BC117076.1 | - | 1 | 587 | 40126395 | ||
BC117080.1 | - | 1 | 587 | 40126399 |