Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164233.1 | NP_001157705.1 | 1 | 180 | ||
NM_027584.1 | NP_081860.1 | 1 | 180 | |||
GenBank mRNA | AK014872.1 | - | 1 | 181 | ||
AK133206.1 | - | 1 | 178 | |||
BC112423.1 | - | 1 | 150 | 40062055 | ||
BC113131.1 | - | 1 | 150 | 40062053 | ||
BY714392.1 | - | 1 | 181 |