Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fance | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163819.1 | NP_001157291.1 | 4 | 1030 | ||
NM_001163820.1 | NP_001157292.1 | 4 | 1030 | |||
GenBank mRNA | AK135017.1 | - | 5 | 934 | ||
AK172283.1 | - | 4 | 1014 | |||
BC016538.1 | - | 4 | 988 | |||
BC039987.1 | - | 4 | 993 |