Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153122.2 | NP_694762.1 | 21 | 3059 | ||
GenBank mRNA | AK164524.1 | - | 5 | 683 | ||
BC025120.1 | - | 8 | 1157 | 4020691 | ||
BC034522.1 | - | 21 | 3059 | 4236444 |