Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sult5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106194.1 | NP_001099664.1 | 5 | 591 | ||
NM_001201369.1 | NP_001188298.1 | 6 | 740 | |||
GenBank mRNA | BF283670.1 | - | 2 | 301 | ||
BQ781676.1 | - | 2 | 320 |