Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019214.1 | NP_062087.1 | 18 | 2176 | ||
XM_008764576.1 | XP_008762798.1 | 19 | 3500 | |||
XM_008764577.1 | XP_008762799.1 | 17 | 2267 | |||
GenBank mRNA | AF167412.1 | - | 18 | 2176 |