Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108245.2 | NP_001101715.1 | 5, 6 | 465 | ||
GenBank mRNA | FM118664.1 | - | 5, 6 | 449 | ||
FQ215476.1 | - | 5, 6 | 460 | |||
FQ216044.1 | - | 5, 6 | 464 | |||
FQ216100.1 | - | 5, 6 | 441 | |||
FQ216185.1 | - | 5, 6 | 461 | |||
FQ224925.1 | - | 5, 6 | 462 |