Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080910.3 | NP_543186.1 | 9 | 1110 | ||
XM_006250852.2 | XP_006250914.1 | 9 | 1158 | |||
XM_006250853.2 | XP_006250915.1 | 9 | 1129 | |||
XM_006250854.2 | XP_006250916.1 | 9 | 1159 | |||
GenBank mRNA | BC072508.1 | - | 9 | 1100 | 7103198 | |
BC085711.1 | - | 9 | 1110 | 7192781 | ||
D37979.1 | - | 7 | 1015 |
Gene Symbol: LOC100910308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751352.3 | XP_003751400.2 | 6 | 886 |