Search
Search

Dnah1| Gene Symbol: Dnah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033655.2 | NP_001028827.2 | 20 | 3486 | ||
| XM_008770977.1 | XP_008769199.1 | 20 | 3544 | |||
| XM_008770978.1 | XP_008769200.1 | 20 | 3544 | |||
| XM_008770979.1 | XP_008769201.1 | 20 | 3544 | |||