Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MKNK1 SIRNA Assays ›
Gene Symbol:
MKNK1
Gene Name:
MAP kinase interacting serine/threonine kinase 1
Gene Aliases:
MNK1
Chromosome Location:
Chr.1: 46557407 - 46607140 on Build GRCh38
UniGene ID:
Hs.371594
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
VHS40573
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MKNK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135553.2
NP_001129025.1
9
843
NM_003684.5
NP_003675.2
10
966
NM_198973.3
NP_945324.1
9
801
NR_024174.2
8
769
NR_024176.2
9
870
XM_006711000.1
XP_006711063.1
9
796
XM_006711001.2
XP_006711064.1
10
1046
XM_006711002.2
XP_006711065.1
9
6271
XM_006711003.1
XP_006711066.1
10
857
XM_006711006.3
XP_006711069.1
8
3586
XM_006711007.1
XP_006711070.1
8
688
XM_006711009.3
XP_006711072.1
8
6173
XM_006711013.1
XP_006711076.1
8
721
XM_006711014.1
XP_006711077.1
8
712
XM_006711015.1
XP_006711078.1
9
783
XM_017002652.1
XP_016858141.1
9
6519
XM_017002653.1
XP_016858142.1
10
1144
XM_017002654.1
XP_016858143.1
10
3996
XM_017002655.1
XP_016858144.1
10
6617
XM_017002656.1
XP_016858145.1
10
6416
XM_017002657.1
XP_016858146.1
9
6318
XM_017002658.1
XP_016858147.1
9
6519
XM_017002659.1
XP_016858148.1
9
815
XM_017002660.1
XP_016858149.1
9
6271
XM_017002661.1
XP_016858150.1
10
1036
XM_017002662.1
XP_016858151.1
10
857
XM_017002663.1
XP_016858152.1
10
6617
XM_017002664.1
XP_016858153.1
8
6197
XM_017002665.1
XP_016858154.1
8
688
XM_017002666.1
XP_016858155.1
9
3672
XM_017002667.1
XP_016858156.1
7
1248
XM_017002668.1
XP_016858157.1
9
6519
GenBank mRNA
AB000409.1
-
9
786
AB451300.1
-
8
599
AB451432.1
-
8
599
AK290456.1
-
9
763
AK298839.1
-
8
743
AK310566.1
-
8
688
AY355461.1
-
8
599
BC002755.2
-
10
886
3629765
BX538193.1
-
7
593
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational initiation
protein phosphorylation
peptidyl-serine phosphorylation
intracellular signal transduction
protein autophosphorylation
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0