Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK8 SIRNA Assays ›
Gene Symbol:
MAPK8
Gene Name:
mitogen-activated protein kinase 8
Gene Aliases:
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
Chromosome Location:
Chr.10: 48306639 - 48439360 on Build GRCh38
UniGene ID:
Hs.138211
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
VHS40724
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278547.1
NP_001265476.1
5
590
NM_001278548.1
NP_001265477.1
4
407
NM_001323302.1
NP_001310231.1
7
853
NM_001323320.1
NP_001310249.1
5
590
NM_001323321.1
NP_001310250.1
5
590
NM_001323322.1
NP_001310251.1
7
853
NM_001323323.1
NP_001310252.1
6
659
NM_001323324.1
NP_001310253.1
5
590
NM_001323325.1
NP_001310254.1
6
784
NM_001323326.1
NP_001310255.1
6
659
NM_001323327.1
NP_001310256.1
6
784
NM_001323328.1
NP_001310257.1
6
659
NM_001323329.1
NP_001310258.1
5
590
NM_001323330.1
NP_001310259.1
6
784
NM_001323331.1
NP_001310260.1
6
784
NM_139046.3
NP_620634.1
7
853
NM_139049.3
NP_620637.1
7
853
NR_136583.1
5
590
NR_136584.1
5
590
NR_136585.1
5
590
GenBank mRNA
AB451231.1
-
4
358
AB451271.1
-
4
358
AB451398.1
-
4
358
AK292523.1
-
5
486
BC130570.1
-
4
420
40146601
BC130572.1
-
4
420
40146603
BC144063.1
-
4
420
9052583
DQ234352.1
-
4
358
L26318.1
-
4
376
U34822.1
-
4
370
U35004.1
-
4
376
U35005.1
-
4
371
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
response to stress
JNK cascade
JUN phosphorylation
response to UV
positive regulation of gene expression
regulation of macroautophagy
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
regulation of histone deacetylation
positive regulation of cyclase activity
negative regulation of protein binding
regulation of protein localization
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
positive regulation of apoptotic process
negative regulation of apoptotic process
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
positive regulation of protein metabolic process
cellular response to lipopolysaccharide
cellular response to mechanical stimulus
positive regulation of deacetylase activity
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
protein serine/threonine kinase activity
JUN kinase activity
protein binding
ATP binding
kinase activity
enzyme binding
histone deacetylase regulator activity
histone deacetylase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-dvzr6:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0