Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PISD-PS3 SIRNA Assays ›
          Gene Symbol:
          Pisd-ps3
          Gene Name:
          phosphatidylserine decarboxylase, pseudogene 3
          Gene Aliases:
          4933439C20Rik
          Chromosome Location:
          Chr.Un|NT_187064: 52674 - 59689 on Build GRCm38
          UniGene ID:
          Mm.247625
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n255837

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pisd-ps3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003518.2 1 68
          GenBank mRNA AK005333.1 - 866
          AK017116.1 - 1 79
          AK034517.1 - 1 726
          AK081907.1 - 1 46
          AK161130.1 - 1 68
          AK162734.1 - 1 1202
          AK168892.1 - 1 46
          AK169597.1 - 1 69
          AK169755.1 - 1 69
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CX207012.1 - 1 68 Chr. Un|NT_187064.1 - 55530 - 59690 [-] on Build GRCm38
          GenBankmRNA CA319555.1 - 12 Chr. Un|NT_187064.1 - 55497 - 59619 [-] on Build GRCm38
          GenBankmRNA AU080934.1 - 85 Chr. - - [-] on Build
          GenBankmRNA BY771791.1 - 1 95 Chr. Un|NT_187064.1 - 55603 - 59716 [-] on Build GRCm38
          GenBankmRNA CJ171901.1 - 1 42 Chr. Un|NT_187064.1 - 57106 - 59663 [-] on Build GRCm38
          GenBankmRNA BY739907.1 - 1 46 Chr. Un|NT_187064.1 - 55575 - 59667 [-] on Build GRCm38
          GenBankmRNA BY255235.1 - 1 46 Chr. Un|NT_187064.1 - 57004 - 59667 [-] on Build GRCm38
          GenBankmRNA BB619177.1 - 1 46 Chr. Un|NT_187064.1 - 55615 - 59667 [-] on Build GRCm38
          GenBankmRNA BQ745548.1 - 1 15 Chr. Un|NT_187064.1 - 55444 - 59637 [-] on Build GRCm38
          GenBankmRNA CJ175330.1 - 1 46 Chr. Un|NT_187064.1 - 57039 - 59667 [-] on Build GRCm38
          GenBankmRNA BY722385.1 - 1 42 Chr. Un|NT_187064.1 - 25366 - 59664 [-] on Build GRCm38
          GenBankmRNA CX237225.1 - 1 116 Chr. Un|NT_187064.1 - 55545 - 59731 [-] on Build GRCm38
          GenBankmRNA BB649681.1 - 1 46 Chr. Un|NT_187064.1 - 55570 - 59667 [-] on Build GRCm38
          GenBankmRNA BY716795.1 - 1 73 Chr. Un|NT_187064.1 - 56853 - 59695 [-] on Build GRCm38
          GenBankmRNA BU611278.1 - 1 183 Chr. Un|NT_187064.1 - 55658 - 59729 [-] on Build GRCm38
          GenBankmRNA CA315346.1 - 1 186 Chr. Un|NT_187064.1 - 55552 - 59792 [-] on Build GRCm38
          GenBankmRNA CJ174094.1 - 1 46 Chr. Un|NT_187064.1 - 57068 - 59667 [-] on Build GRCm38
          GenBankmRNA BY771278.1 - 1 95 Chr. Un|NT_187064.1 - 55642 - 59716 [-] on Build GRCm38
          GenBankmRNA BY273730.1 - 1 47 Chr. Un|NT_187064.1 - 57051 - 59667 [-] on Build GRCm38
          GenBankmRNA BY790085.1 - 1 95 Chr. Un|NT_187064.1 - 55628 - 59716 [-] on Build GRCm38
          GenBankmRNA AU066585.1 - 1 67 Chr. 11 - 3124028 - 3127613 [+] on Build GRCm38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          decarboxylase

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dt4l6:80/100.66.76.4:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0