Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL4 SIRNA Assays ›
          Gene Symbol:
          Il4
          Gene Name:
          interleukin 4
          Gene Aliases:
          BSF-1, Il-4
          Chromosome Location:
          Chr.11: 53612460 - 53618665 on Build GRCm38
          UniGene ID:
          Mm.276360
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n257566

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Il4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027491.1 1 222
          GenBank mRNA AA199406.1 - 1 222
          AF359558.1 - 1 161
          BX529138.1 - 1 231
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AI324371.1 - 1 60 Chr. 11 - 53612469 - 53617176 [-] on Build GRCm38
          GenBankmRNA AA153220.1 - 1 60 Chr. 11 - 53612474 - 53617176 [-] on Build GRCm38
          GenBankmRNA AW988069.1 - 1 219 Chr. 11 - 53612468 - 53617335 [-] on Build GRCm38
          GenBankmRNA AA967539.1 - 222 Chr. - - [-] on Build

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of protein phosphorylation
          innate immune response in mucosa
          positive regulation of defense response to virus by host
          T-helper 1 cell lineage commitment
          positive regulation of immunoglobulin production
          negative regulation of acute inflammatory response
          negative regulation of chronic inflammatory response
          immune response
          cholesterol metabolic process
          positive regulation of cell proliferation
          regulation of proton transport
          negative regulation of epithelial cell migration
          positive regulation of B cell proliferation
          B cell costimulation
          positive regulation of interleukin-10 production
          positive regulation of interleukin-13 production
          T-helper 2 cell cytokine production
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          B cell activation
          positive regulation of tyrosine phosphorylation of Stat5 protein
          defense response to protozoan
          negative regulation of macrophage activation
          positive regulation of mast cell degranulation
          negative regulation of nitric oxide biosynthetic process
          T-helper 2 cell differentiation
          positive regulation of chemokine biosynthetic process
          positive regulation of MHC class II biosynthetic process
          positive regulation of T cell differentiation
          negative regulation of osteoclast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of isotype switching to IgE isotypes
          positive regulation of isotype switching to IgG isotypes
          regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of immune response
          negative regulation of T cell activation
          positive regulation of B cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of mononuclear cell migration
          dendritic cell differentiation
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of myoblast fusion
          negative regulation of complement-dependent cytotoxicity
          negative regulation of T-helper 17 cell differentiation
          negative regulation of endothelial cell apoptotic process
          positive regulation of eosinophil chemotaxis
          negative regulation of extrinsic apoptotic signaling pathway
          cytokine activity
          cytokine receptor binding
          interleukin-4 receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0