Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYP2B7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001278.1 | 1 | 79 | |||
GenBank mRNA | AK307933.1 | - | 1 | 125 | ||
DQ198366.1 | - | 1 | 71 | |||
DQ198367.1 | - | 1 | 71 |