Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GBAP1 SIRNA Assays ›
          Gene Symbol:
          GBAP1
          Gene Name:
          glucosylceramidase beta pseudogene 1
          Gene Aliases:
          GBAP
          Chromosome Location:
          Chr.1: 155213825 - 155227534 on Build GRCh38
          UniGene ID:
          Hs.719930
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n262039

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GBAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002188.3 3 265
          GenBank mRNA AK295259.1 - 2 175
          BC030240.1 - 3 144
          BC092482.1 - 2 175
          DB192327.1 - 3 246
          KJ690774.1 - 2 93
          KJ690775.1 - 2 93
          KJ690776.1 - 2 93
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA080524.1 - 2 174 Chr. 1 - 155218067 - 155219004 [-] on Build GRCh38
          GenBankmRNA DA233497.1 - 2 174 Chr. 1 - 155218062 - 155219004 [-] on Build GRCh38
          GenBankmRNA DA449111.1 - 3 220 Chr. 1 - 155218119 - 155228016 [-] on Build GRCh38
          GenBankmRNA DA317149.1 - 2 178 Chr. 1 - 155218096 - 155219008 [-] on Build GRCh38
          GenBankmRNA DA224838.1 - 2 162 Chr. 1 - 155218058 - 155218993 [-] on Build GRCh38
          GenBankmRNA DB253703.1 - 2 175 Chr. 1 - 155218152 - 155219005 [-] on Build GRCh38
          GenBankmRNA DA068957.1 - 2 186 Chr. 1 - 155218074 - 155219016 [-] on Build GRCh38
          GenBankmRNA DA345905.1 - 3 202 Chr. 1 - 155218075 - 155227469 [-] on Build GRCh38
          GenBankmRNA DA353838.1 - 2 165 Chr. 1 - 155218055 - 155218995 [-] on Build GRCh38
          GenBankmRNA DA397642.1 - 2 175 Chr. 1 - 155218046 - 155219005 [-] on Build GRCh38
          GenBankmRNA HY139750.1 - 2 192 Chr. 1 - 155218184 - 155219022 [-] on Build GRCh38
          GenBankmRNA DA219155.1 - 2 158 Chr. 1 - 155218049 - 155218988 [-] on Build GRCh38
          GenBankmRNA BI256842.1 - 3 133 Chr. 1 - 155216429 - 155244526 [-] on Build GRCh38
          GenBankmRNA DA532484.1 - 2 181 Chr. 1 - 155218175 - 155219011 [-] on Build GRCh38
          GenBankmRNA DA164501.1 - 2 187 Chr. 1 - 155218084 - 155219017 [-] on Build GRCh38
          GenBankmRNA DA233555.1 - 2 192 Chr. 1 - 155218065 - 155219022 [-] on Build GRCh38
          GenBankmRNA BQ218826.1 - 3 269 Chr. 1 - 155217920 - 155227522 [-] on Build GRCh38
          GenBankmRNA DA188934.1 - 2 200 Chr. 1 - 155218120 - 155219028 [-] on Build GRCh38
          GenBankmRNA DA057140.1 - 2 167 Chr. 1 - 155218051 - 155218997 [-] on Build GRCh38
          GenBankmRNA DA224846.1 - 2 210 Chr. 1 - 155218136 - 155219040 [-] on Build GRCh38
          GenBankmRNA DA769889.1 - 3 161 Chr. 1 - 155218071 - 155227430 [-] on Build GRCh38
          GenBankmRNA DA219124.1 - 2 158 Chr. 1 - 155218051 - 155218988 [-] on Build GRCh38
          GenBankmRNA DA107001.1 - 3 267 Chr. 1 - 155218155 - 155227533 [-] on Build GRCh38
          GenBankmRNA DA179266.1 - 2 169 Chr. 1 - 155218169 - 155218999 [-] on Build GRCh38
          GenBankmRNA BI770380.1 - 3 113 Chr. 1 - 155217738 - 155244525 [-] on Build GRCh38
          GenBankmRNA DA217255.1 - 2 193 Chr. 1 - 155218108 - 155219021 [-] on Build GRCh38
          GenBankmRNA DA124268.1 - 2 175 Chr. 1 - 155213464 - 155219005 [-] on Build GRCh38
          GenBankmRNA DA236364.1 - 2 158 Chr. 1 - 155218083 - 155218988 [-] on Build GRCh38
          GenBankmRNA DA798350.1 - 2 203 Chr. 1 - 155218108 - 155219033 [-] on Build GRCh38
          GenBankmRNA DA876012.1 - 2 192 Chr. 1 - 155218088 - 155219022 [-] on Build GRCh38
          GenBankmRNA DA338971.1 - 2 158 Chr. 1 - 155218037 - 155218988 [-] on Build GRCh38
          GenBankmRNA DA525119.1 - 3 159 Chr. 1 - 155218063 - 155227428 [-] on Build GRCh38
          GenBankmRNA DA221157.1 - 2 175 Chr. 1 - 155218115 - 155219004 [-] on Build GRCh38
          GenBankmRNA DA678288.1 - 3 104 Chr. 1 - 155216493 - 155227428 [-] on Build GRCh38
          GenBankmRNA DA096069.1 - 3 270 Chr. 1 - 155218177 - 155227539 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lv5mj:80/100.66.78.219:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0