Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024616.1 | 1 | 636 | |||
GenBank mRNA | BC069671.1 | - | 1 | 559 | ||
BC069696.1 | - | 1 | 558 | |||
BC069772.1 | - | 1 | 558 | |||
Y10696.1 | - | 1 | 562 |