Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG2 SIRNA Assays ›
          Gene Symbol:
          ALG2
          Gene Name:
          ALG2, alpha-1,3/1,6-mannosyltransferase
          Gene Aliases:
          CDGIi, CMS14, CMSTA3, NET38, hALPG2
          Chromosome Location:
          Chr.9: 99216425 - 99221964 on Build GRCh38
          UniGene ID:
          Hs.40919
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263334

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024532.1 2 479
          GenBank mRNA AK074988.1 - 2 457
          AY358697.1 - 1 64
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AI768004.1 - 2 122 Chr. 9 - 99218594 - 99221607 [-] on Build GRCh38
          GenBankmRNA AW051481.1 - 2 122 Chr. 9 - 99218621 - 99221607 [-] on Build GRCh38
          GenBankmRNA CF132606.1 - 2 246 Chr. 9 - 99218712 - 99221731 [-] on Build GRCh38
          GenBankmRNA DA890710.1 - 2 456 Chr. 9 - 99221011 - 99221940 [-] on Build GRCh38
          GenBankmRNA BP273835.1 - 2 427 Chr. 9 - 99220978 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA430466.1 - 2 428 Chr. 9 - 99218796 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA422820.1 - 2 449 Chr. 9 - 99218808 - 99221923 [-] on Build GRCh38
          GenBankmRNA DB059946.1 - 2 427 Chr. 9 - 99221029 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA736744.1 - 2 457 Chr. 9 - 99218649 - 99221942 [-] on Build GRCh38
          GenBankmRNA BQ641538.1 - 369 Chr. - - [-] on Build
          GenBankmRNA BU594798.1 - 2 460 Chr. 9 - 99218625 - 99221944 [-] on Build GRCh38
          GenBankmRNA DB460341.1 - 2 427 Chr. 9 - 99221049 - 99221912 [-] on Build GRCh38
          GenBankmRNA HY019575.1 - 2 189 Chr. 9 - 99218635 - 99221548 [-] on Build GRCh38
          GenBankmRNA DB187551.1 - 2 432 Chr. 9 - 99218789 - 99221912 [-] on Build GRCh38
          GenBankmRNA AI278375.1 - 2 122 Chr. 9 - 99218609 - 99221607 [-] on Build GRCh38
          GenBankmRNA DA627469.1 - 2 455 Chr. 9 - 99220989 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA714128.1 - 2 437 Chr. 9 - 99218795 - 99221914 [-] on Build GRCh38
          GenBankmRNA DA250901.1 - 2 455 Chr. 9 - 99218811 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA666759.1 - 2 457 Chr. 9 - 99220988 - 99221942 [-] on Build GRCh38
          GenBankmRNA BP342204.1 - 2 427 Chr. 9 - 99218778 - 99221912 [-] on Build GRCh38
          GenBankmRNA DA928356.1 - 2 435 Chr. 9 - 99218812 - 99221916 [-] on Build GRCh38
          GenBankmRNA DB123779.1 - 2 455 Chr. 9 - 99218812 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA439254.1 - 2 427 Chr. 9 - 99218817 - 99221912 [-] on Build GRCh38
          GenBankmRNA DB195604.1 - 2 441 Chr. 9 - 99221005 - 99221926 [-] on Build GRCh38
          GenBankmRNA CF131378.1 - 2 278 Chr. 9 - 99218633 - 99221763 [-] on Build GRCh38
          GenBankmRNA DA904634.1 - 2 456 Chr. 9 - 99220977 - 99221940 [-] on Build GRCh38
          GenBankmRNA DA862318.1 - 2 439 Chr. 9 - 99221029 - 99221916 [-] on Build GRCh38
          GenBankmRNA AI863884.1 - 2 122 Chr. 9 - 99218654 - 99221607 [-] on Build GRCh38
          GenBankmRNA DR006890.1 - 2 336 Chr. 9 - 99218507 - 99221816 [-] on Build GRCh38
          GenBankmRNA BG773528.1 - 2 274 Chr. 9 - 99218494 - 99221523 [-] on Build GRCh38
          GenBankmRNA AA769683.1 - 2 121 Chr. 9 - 99218552 - 99221606 [-] on Build GRCh38
          GenBankmRNA DA608501.1 - 2 457 Chr. 9 - 99220999 - 99221942 [-] on Build GRCh38
          GenBankmRNA DB000389.1 - 2 456 Chr. 9 - 99220998 - 99221940 [-] on Build GRCh38
          GenBankmRNA AI183323.1 - 2 122 Chr. 9 - 99218659 - 99221607 [-] on Build GRCh38
          GenBankmRNA DA371411.1 - 2 427 Chr. 9 - 99218812 - 99221912 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          dolichol-linked oligosaccharide biosynthetic process
          protein glycosylation in endoplasmic reticulum
          response to calcium ion
          mannosylation
          alpha-1,3-mannosyltransferase activity
          GDP-Man:Man1GlcNAc2-PP-Dol alpha-1,3-mannosyltransferase activity
          protein binding
          protein heterodimerization activity
          protein N-terminus binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0