Search
Search

MOBP| Gene Symbol: MOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003090.2 | 6 | 1340 | |||
| NR_103504.1 | 7 | 1682 | ||||
| NR_103505.1 | 7 | 1720 | ||||
| GenBank mRNA | AK095180.1 | - | 2 | 2627 | ||
| BC022471.1 | - | 6 | 1235 | |||
| BC119694.1 | - | 5 | 1117 | 40116789 | ||
| BC119695.1 | - | 5 | 1118 | 40116791 | ||