Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUNX1-IT1 SIRNA Assays ›
          Gene Symbol:
          RUNX1-IT1
          Gene Name:
          RUNX1 intronic transcript 1
          Gene Aliases:
          C21orf96
          Chromosome Location:
          Chr.21: 35037925 - 35039426 on Build GRCh38
          UniGene ID:
          Hs.672131
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n268598

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUNX1-IT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026812.1 1 1373
          GenBank mRNA AK024509.1 - 1 1367

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline