Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR2F1-AS1 SIRNA Assays ›
          Gene Symbol:
          NR2F1-AS1
          Gene Name:
          NR2F1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 93409356 - 93581320 on Build GRCh38
          UniGene ID:
          Hs.457407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269416

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR2F1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_021490.2 4 446
          NR_021491.2 4 446
          NR_109818.1 5 559
          NR_109819.1 4 1121
          NR_109820.1 4 446
          NR_109821.1 4 446
          NR_109823.1 4 443
          NR_109824.1 4 446
          NR_109825.1 5 1226
          GenBank mRNA AK095719.1 - 5 537
          AK124699.1 - 4 1171
          AK311323.1 - 4 834
          BC010610.1 - 4 414
          BC018089.1 - 4 423
          BC042879.1 - 4 345
          BC044619.1 - 5 702
          BC048999.1 - 5 702
          BC144331.1 - 4 1721
          BX647510.1 - 3 214
          DA718791.1 - 4 429
          DB257894.1 - 4 345
          HY108556.1 - 4 376
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB503335.1 - 4 342 Chr. 5 - 93544834 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA531679.1 - 4 497 Chr. 5 - 93543968 - 93581370 [-] on Build GRCh38
          GenBankmRNA BM546601.1 - 511 Chr. - - [-] on Build
          GenBankmRNA DA271332.1 - 4 360 Chr. 5 - 93438714 - 93581234 [-] on Build GRCh38
          GenBankmRNA HY136165.1 - 5 403 Chr. 5 - 93553771 - 93581276 [-] on Build GRCh38
          GenBankmRNA DB486010.2 - 4 342 Chr. 5 - 93411386 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA533837.1 - 4 419 Chr. 5 - 93544815 - 93580618 [-] on Build GRCh38
          GenBankmRNA DA363045.1 - 4 490 Chr. 5 - 93553758 - 93580689 [-] on Build GRCh38
          GenBankmRNA DA527197.1 - 4 429 Chr. 5 - 93411404 - 93581303 [-] on Build GRCh38
          GenBankmRNA DA706940.1 - 4 428 Chr. 5 - 93544828 - 93581300 [-] on Build GRCh38
          GenBankmRNA DA280798.1 - 4 327 Chr. 5 - 93411352 - 93581199 [-] on Build GRCh38
          GenBankmRNA BX377086.2 - 4 349 Chr. 5 - 93543540 - 93581223 [-] on Build GRCh38
          GenBankmRNA BG701915.1 - 4 589 Chr. 5 - 93501042 - 93585650 [-] on Build GRCh38
          GenBankmRNA DA844094.1 - 4 425 Chr. 5 - 93544836 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA066475.1 - 2 110 Chr. 5 - 93411149 - 93554938 [-] on Build GRCh38
          GenBankmRNA BI669922.1 - 4 350 Chr. 5 - 93411232 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA518837.1 - 5 537 Chr. 5 - 93553785 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA545967.1 - 4 345 Chr. 5 - 93544816 - 93581219 [-] on Build GRCh38
          GenBankmRNA DB272535.1 - 4 447 Chr. 5 - 93544853 - 93580646 [-] on Build GRCh38
          GenBankmRNA BG715542.1 - 4 430 Chr. 5 - 93411271 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA304607.1 - 4 332 Chr. 5 - 93411321 - 93581205 [-] on Build GRCh38
          GenBankmRNA DB498974.1 - 4 342 Chr. 5 - 93544802 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA839566.1 - 3 236 Chr. 5 - 93432362 - 93563432 [-] on Build GRCh38
          GenBankmRNA HY141502.1 - 4 326 Chr. 5 - 93544815 - 93581199 [-] on Build GRCh38
          GenBankmRNA BG707159.1 - 4 350 Chr. 5 - 93411365 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA388648.1 - 4 496 Chr. 5 - 93544889 - 93580695 [-] on Build GRCh38
          GenBankmRNA DA328801.1 - 4 347 Chr. 5 - 93543992 - 93581221 [-] on Build GRCh38
          GenBankmRNA BI820847.1 - 4 396 Chr. 5 - 93342240 - 93580595 [-] on Build GRCh38
          GenBankmRNA DA517345.1 - 4 422 Chr. 5 - 93411434 - 93581295 [-] on Build GRCh38
          GenBankmRNA DB492381.1 - 4 342 Chr. 5 - 93544849 - 93581216 [-] on Build GRCh38
          GenBankmRNA DB475119.2 - 4 420 Chr. 5 - 93411324 - 93581297 [-] on Build GRCh38
          GenBankmRNA DB177914.1 - 4 426 Chr. 5 - 93438700 - 93581300 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-v49qv:80/100.66.72.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0