Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR2F1-AS1 SIRNA Assays ›
          Gene Symbol:
          NR2F1-AS1
          Gene Name:
          NR2F1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 93409356 - 93581320 on Build GRCh38
          UniGene ID:
          Hs.457407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269418

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR2F1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_021490.2 3 340
          NR_021491.2 3 340
          NR_109818.1 4 453
          NR_109819.1 3 1015
          NR_109820.1 3 340
          NR_109821.1 3 340
          NR_109823.1 3 337
          NR_109824.1 3 340
          NR_109825.1 4 1120
          GenBank mRNA AK095719.1 - 4 431
          AK124699.1 - 3 1065
          AK311143.1 - 3 239
          AK311323.1 - 3 728
          BC010610.1 - 3 308
          BC018089.1 - 3 317
          BC042879.1 - 3 239
          BC044619.1 - 4 596
          BC048999.1 - 4 596
          BC144330.1 - 3 1615
          BC144331.1 - 3 1615
          BX647510.1 - 2 108
          DA718791.1 - 3 323
          DB257894.1 - 3 239
          HY108556.1 - 3 270
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB503335.1 - 3 236 Chr. 5 - 93544834 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA531679.1 - 3 391 Chr. 5 - 93543968 - 93581370 [-] on Build GRCh38
          GenBankmRNA BI463026.1 - 4 603 Chr. 5 - 93554872 - 93570826 [-] on Build GRCh38
          GenBankmRNA DB045052.1 - 4 530 Chr. 5 - 93554901 - 93570793 [-] on Build GRCh38
          GenBankmRNA BM546601.1 - 405 Chr. - - [-] on Build
          GenBankmRNA DA271332.1 - 3 254 Chr. 5 - 93438714 - 93581234 [-] on Build GRCh38
          GenBankmRNA HY136165.1 - 4 297 Chr. 5 - 93553771 - 93581276 [-] on Build GRCh38
          GenBankmRNA DB173308.1 - 3 490 Chr. 5 - 93553837 - 93580795 [-] on Build GRCh38
          GenBankmRNA DB486010.2 - 3 236 Chr. 5 - 93411386 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA533837.1 - 3 313 Chr. 5 - 93544815 - 93580618 [-] on Build GRCh38
          GenBankmRNA DA363045.1 - 3 384 Chr. 5 - 93553758 - 93580689 [-] on Build GRCh38
          GenBankmRNA BF530372.1 - 312 Chr. - - [-] on Build
          GenBankmRNA DA134670.1 - 3 239 Chr. 5 - 93411404 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA527197.1 - 3 323 Chr. 5 - 93411404 - 93581303 [-] on Build GRCh38
          GenBankmRNA DA706940.1 - 3 322 Chr. 5 - 93544828 - 93581300 [-] on Build GRCh38
          GenBankmRNA DA280798.1 - 3 221 Chr. 5 - 93411352 - 93581199 [-] on Build GRCh38
          GenBankmRNA BX377086.2 - 3 243 Chr. 5 - 93543540 - 93581223 [-] on Build GRCh38
          GenBankmRNA BG701915.1 - 3 484 Chr. 5 - 93501042 - 93585650 [-] on Build GRCh38
          GenBankmRNA DA844094.1 - 3 319 Chr. 5 - 93544836 - 93581297 [-] on Build GRCh38
          GenBankmRNA AA375334.1 - 3 188 Chr. 5 - 93553829 - 93581168 [-] on Build GRCh38
          GenBankmRNA BI669922.1 - 3 244 Chr. 5 - 93411232 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA545967.1 - 3 239 Chr. 5 - 93544816 - 93581219 [-] on Build GRCh38
          GenBankmRNA DB272535.1 - 3 341 Chr. 5 - 93544853 - 93580646 [-] on Build GRCh38
          GenBankmRNA DA518837.1 - 4 431 Chr. 5 - 93553785 - 93581297 [-] on Build GRCh38
          GenBankmRNA BG715542.1 - 3 324 Chr. 5 - 93411271 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA304607.1 - 3 226 Chr. 5 - 93411321 - 93581205 [-] on Build GRCh38
          GenBankmRNA DB498974.1 - 3 236 Chr. 5 - 93544802 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA839566.1 - 2 130 Chr. 5 - 93432362 - 93563432 [-] on Build GRCh38
          GenBankmRNA HY141502.1 - 3 220 Chr. 5 - 93544815 - 93581199 [-] on Build GRCh38
          GenBankmRNA BG707159.1 - 3 244 Chr. 5 - 93411365 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA328801.1 - 3 241 Chr. 5 - 93543992 - 93581221 [-] on Build GRCh38
          GenBankmRNA DA388648.1 - 3 390 Chr. 5 - 93544889 - 93580695 [-] on Build GRCh38
          GenBankmRNA DN993098.1 - 3 441 Chr. 5 - 93553814 - 93580737 [-] on Build GRCh38
          GenBankmRNA BI820847.1 - 3 290 Chr. 5 - 93342240 - 93580595 [-] on Build GRCh38
          GenBankmRNA DA517345.1 - 3 316 Chr. 5 - 93411434 - 93581295 [-] on Build GRCh38
          GenBankmRNA DB492381.1 - 3 236 Chr. 5 - 93544849 - 93581216 [-] on Build GRCh38
          GenBankmRNA DB475119.2 - 3 314 Chr. 5 - 93411324 - 93581297 [-] on Build GRCh38
          GenBankmRNA DB177914.1 - 3 320 Chr. 5 - 93438700 - 93581300 [-] on Build GRCh38
          GenBankmRNA DB030965.1 - 4 499 Chr. 5 - 93554880 - 93570722 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2lcrp:80/100.66.72.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0