Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUTM2A-AS1 SIRNA Assays ›
          Gene Symbol:
          NUTM2A-AS1 NUTM2B-AS1
          Gene Name:
          NUTM2A antisense RNA 1
          NUTM2B antisense RNA 1
          Gene Aliases:
          FAM22A-AS1
          -
          Chromosome Location:
          Chr.10: 87238667 - 87342558 on Build GRCh38
          Chromosome Location:
          Chr.10: 79691500 - 79826594 on Build GRCh38
          UniGene ID:
          Hs.440648
          Hs.440648
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n270292

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUTM2A-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024397.1 2 366
          GenBank mRNA AK309777.1 - 2 185
          AK311174.1 - 2 205
          BC047063.1 - 2 293
          DB257428.1 - 2 367
          Gene Symbol: NUTM2B-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_120611.1 2 446
          NR_120612.1 2 446
          NR_120613.1 2 446
          GenBank mRNA BQ425131.1 - 2 466
          CB163469.1 - 2 263
          CN279211.1 - 2 306
          CN279213.1 - 2 290
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA040049.1 - 2 185 Chr. 10 - 87245755 - 87342377 [-] on Build GRCh38
          GenBankmRNA DA737563.1 - 2 316 Chr. 10 - 87245656 - 87342506 [-] on Build GRCh38
          GenBankmRNA BI767196.1 - 174 Chr. - - [-] on Build
          GenBankmRNA DA796822.1 - 2 350 Chr. 10 - 87288387 - 87342541 [-] on Build GRCh38
          GenBankmRNA AL520958.3 - 2 89 Chr. 10 - 79797377 - 79826237 [-] on Build GRCh38
          GenBankmRNA DA691562.1 - 2 329 Chr. 10 - 79804494 - 79826468 [-] on Build GRCh38
          GenBankmRNA BG111204.1 - 2 107 Chr. 10 - 79691671 - 79826255 [-] on Build GRCh38
          GenBankmRNA DA010830.1 - 3 380 Chr. 10 - 79806056 - 79826563 [-] on Build GRCh38
          GenBankmRNA BG166579.1 - 2 345 Chr. 10 - 79766137 - 79826484 [-] on Build GRCh38
          GenBankmRNA CD250242.1 - 2 373 Chr. 10 - 87288178 - 87342541 [-] on Build GRCh38
          GenBankmRNA HY154971.1 - 2 294 Chr. 10 - 87216096 - 87342485 [-] on Build GRCh38
          GenBankmRNA CN274008.1 - 2 178 Chr. 10 - 79802853 - 79826326 [-] on Build GRCh38
          GenBankmRNA BM559658.1 - 282 Chr. - - [-] on Build
          GenBankmRNA HY062022.1 - 2 242 Chr. 10 - 87288324 - 87342412 [-] on Build GRCh38
          GenBankmRNA DB483325.1 - 2 264 Chr. 10 - 87246167 - 87342456 [-] on Build GRCh38
          GenBankmRNA BG437437.1 - 414 Chr. - - [-] on Build
          GenBankmRNA HY087386.1 - 2 242 Chr. 10 - 87288299 - 87342412 [-] on Build GRCh38
          GenBankmRNA DA200463.1 - 2 205 Chr. 10 - 87206423 - 87342397 [-] on Build GRCh38
          GenBankmRNA CN279215.1 - 2 330 Chr. 10 - 79804484 - 79826470 [-] on Build GRCh38
          GenBankmRNA AW612231.1 - 2 244 Chr. 10 - 79804492 - 79826392 [-] on Build GRCh38
          GenBankmRNA HY150000.1 - 2 184 Chr. 10 - 87216123 - 87342375 [-] on Build GRCh38
          GenBankmRNA T58826.1 - 101 Chr. - - [-] on Build
          GenBankmRNA HY010169.1 - 2 264 Chr. 10 - 87238758 - 87342455 [-] on Build GRCh38
          GenBankmRNA HY124799.1 - 2 156 Chr. 10 - 87216064 - 87342347 [-] on Build GRCh38
          GenBankmRNA BG748770.1 - 2 368 Chr. 10 - 79766290 - 79826477 [-] on Build GRCh38
          GenBankmRNA BE742363.1 - 263 Chr. - - [-] on Build
          GenBankmRNA DB456680.1 - 2 293 Chr. 10 - 87246163 - 87342485 [-] on Build GRCh38
          GenBankmRNA HY151352.1 - 2 286 Chr. 10 - 87208482 - 87342477 [-] on Build GRCh38
          GenBankmRNA DA829196.1 - 2 163 Chr. 10 - 87287993 - 87342355 [-] on Build GRCh38
          GenBankmRNA DA571282.1 - 2 208 Chr. 10 - 87288087 - 87342400 [-] on Build GRCh38
          GenBankmRNA DA258253.1 - 2 185 Chr. 10 - 87216011 - 87342377 [-] on Build GRCh38
          GenBankmRNA DA259512.1 - 2 191 Chr. 10 - 87202973 - 87342383 [-] on Build GRCh38
          GenBankmRNA DA551755.1 - 2 151 Chr. 10 - 87245749 - 87342343 [-] on Build GRCh38
          GenBankmRNA BG717953.1 - 2 381 Chr. 10 - 79691997 - 79826516 [-] on Build GRCh38
          GenBankmRNA DB263965.1 - 2 178 Chr. 10 - 87206378 - 87342370 [-] on Build GRCh38
          GenBankmRNA HY055033.1 - 2 264 Chr. 10 - 87288436 - 87342455 [-] on Build GRCh38
          GenBankmRNA DA009741.1 - 2 183 Chr. 10 - 87245758 - 87342375 [-] on Build GRCh38
          GenBankmRNA BM053142.1 - 2 376 Chr. 10 - 79808759 - 79826515 [-] on Build GRCh38
          GenBankmRNA AW204120.1 - 2 243 Chr. 10 - 79804494 - 79826391 [-] on Build GRCh38
          GenBankmRNA DA775058.1 - 2 186 Chr. 10 - 87215939 - 87342377 [-] on Build GRCh38
          GenBankmRNA HY157097.1 - 2 156 Chr. 10 - 87216111 - 87342347 [-] on Build GRCh38
          GenBankmRNA BM456581.1 - 589 Chr. - - [-] on Build
          GenBankmRNA BG256210.1 - 235 Chr. - - [-] on Build
          GenBankmRNA CN279212.1 - 2 288 Chr. 10 - 79804510 - 79826427 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dg9q5:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0