Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG15 SIRNA Assays ›
          Gene Symbol:
          SNHG15
          Gene Name:
          small nucleolar RNA host gene 15
          Gene Aliases:
          C7orf40, Linc-Myo1g, MYO1GUT
          Chromosome Location:
          Chr.7: 44983028 - 44986660 on Build GRCh38
          UniGene ID:
          Hs.25892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270572

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003697.1 2, 3 221
          GenBank mRNA BC092459.1 - 2, 3 221
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AA524713.1 - 2, 3 313 Chr. 7 - 44983904 - 44986757 [-] on Build GRCh38
          GenBankmRNA BE889371.1 - 2, 3 127 Chr. 7 - 44983041 - 44986566 [-] on Build GRCh38
          GenBankmRNA R70423.1 - 187 Chr. - - [-] on Build
          GenBankmRNA W57948.1 - 192 Chr. - - [-] on Build
          GenBankmRNA BM710323.1 - 2, 3 242 Chr. 7 - 44983870 - 44986681 [-] on Build GRCh38
          GenBankmRNA HX788853.1 - 2, 3 210 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BE888491.1 - 165 Chr. - - [-] on Build
          GenBankmRNA BF304363.1 - 147 Chr. - - [-] on Build
          GenBankmRNA BE206230.1 - 2, 3 122 Chr. 7 - 44983229 - 44986559 [-] on Build GRCh38
          GenBankmRNA AA366086.1 - 2, 3 200 Chr. 7 - 44983993 - 44986638 [-] on Build GRCh38
          GenBankmRNA BF131634.1 - 161 Chr. - - [-] on Build
          GenBankmRNA BF306650.1 - 150 Chr. - - [-] on Build
          GenBankmRNA AW245923.1 - 2, 3 226 Chr. 7 - 44983229 - 44986640 [-] on Build GRCh38
          GenBankmRNA BF204739.1 - 2, 3 220 Chr. 7 - 44983092 - 44986658 [-] on Build GRCh38
          GenBankmRNA BU195547.1 - 2, 3 169 Chr. 7 - 44983030 - 44986605 [-] on Build GRCh38
          GenBankmRNA DB490500.1 - 2, 3 221 Chr. 7 - 44984357 - 44986660 [-] on Build GRCh38
          GenBankmRNA BG491747.1 - 2, 3 243 Chr. 7 - 44983061 - 44986681 [-] on Build GRCh38
          GenBankmRNA CF128961.1 - 2, 3 186 Chr. 7 - 44983832 - 44986625 [-] on Build GRCh38
          GenBankmRNA BU502253.1 - 2, 3 168 Chr. 7 - 44983028 - 44986604 [-] on Build GRCh38
          GenBankmRNA BE733369.1 - 2, 3 395 Chr. 7 - 44983106 - 44986834 [-] on Build GRCh38
          GenBankmRNA BM744843.1 - 2, 3 185 Chr. 7 - 44983285 - 44986617 [-] on Build GRCh38
          GenBankmRNA BG677227.1 - 2, 3 158 Chr. 7 - 44983131 - 44986597 [-] on Build GRCh38
          GenBankmRNA CD640711.1 - 2, 3 171 Chr. 7 - 44983119 - 44986607 [-] on Build GRCh38
          GenBankmRNA BF314122.1 - 2, 3 147 Chr. 7 - 44983046 - 44986586 [-] on Build GRCh38
          GenBankmRNA BG829241.1 - 2, 3 220 Chr. 7 - 44983237 - 44986619 [-] on Build GRCh38
          GenBankmRNA BG927882.1 - 2, 3 263 Chr. 7 - 44983170 - 44986619 [-] on Build GRCh38
          GenBankmRNA AA252002.1 - 2, 3 198 Chr. 7 - 44983817 - 44986638 [-] on Build GRCh38
          GenBankmRNA BM772795.1 - 2, 3 185 Chr. 7 - 44983206 - 44986617 [-] on Build GRCh38
          GenBankmRNA BM758831.1 - 2, 3 185 Chr. 7 - 44983894 - 44986624 [-] on Build GRCh38
          GenBankmRNA DB484794.1 - 2, 3 221 Chr. 7 - 44983890 - 44986660 [-] on Build GRCh38
          GenBankmRNA BM685499.1 - 2, 3 179 Chr. 7 - 44983169 - 44986611 [-] on Build GRCh38
          GenBankmRNA AB371433.1 - 2, 3 199 Chr. 7 - 44983029 - 44986637 [-] on Build GRCh38
          GenBankmRNA BU537835.1 - 2, 3 229 Chr. 7 - 44983141 - 44986668 [-] on Build GRCh38
          GenBankmRNA AV759018.1 - 2, 3 225 Chr. 7 - 44983831 - 44986664 [-] on Build GRCh38
          GenBankmRNA BY996199.1 - 2, 3 175 Chr. 7 - 44983146 - 44986608 [-] on Build GRCh38
          GenBankmRNA AW960033.1 - 2, 3 198 Chr. 7 - 44983173 - 44986637 [-] on Build GRCh38
          GenBankmRNA AW328480.1 - 2, 3 136 Chr. 7 - 44983242 - 44986566 [-] on Build GRCh38
          GenBankmRNA BQ059848.1 - 139 Chr. - - [-] on Build
          GenBankmRNA BM887994.1 - 2, 3 149 Chr. 7 - 44983268 - 44986588 [-] on Build GRCh38
          GenBankmRNA BG829322.1 - 2, 3 220 Chr. 7 - 44983089 - 44986658 [-] on Build GRCh38
          GenBankmRNA DB492530.1 - 2, 3 222 Chr. 7 - 44983886 - 44986660 [-] on Build GRCh38
          GenBankmRNA BM764510.1 - 2, 3 185 Chr. 7 - 44983184 - 44986617 [-] on Build GRCh38
          GenBankmRNA N35269.1 - 2, 3 180 Chr. 7 - 44983929 - 44986621 [-] on Build GRCh38
          GenBankmRNA BE312499.1 - 2, 3 127 Chr. 7 - 44983106 - 44986566 [-] on Build GRCh38
          GenBankmRNA BM773240.1 - 2, 3 185 Chr. 7 - 44983897 - 44986617 [-] on Build GRCh38
          GenBankmRNA AA258914.1 - 2, 3 169 Chr. 7 - 44983817 - 44986598 [-] on Build GRCh38
          GenBankmRNA HX785737.1 - 2, 3 210 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BG748946.1 - 2, 3 186 Chr. 7 - 44983065 - 44986624 [-] on Build GRCh38
          GenBankmRNA AA306794.1 - 2, 3 156 Chr. 7 - 44983244 - 44986595 [-] on Build GRCh38
          GenBankmRNA BG548028.1 - 2, 3 227 Chr. 7 - 44983041 - 44986665 [-] on Build GRCh38
          GenBankmRNA BQ434731.1 - 2, 3 199 Chr. 7 - 44983024 - 44986638 [-] on Build GRCh38
          GenBankmRNA CD522132.1 - 224 Chr. - - [-] on Build
          GenBankmRNA AA281810.1 - 2, 3 198 Chr. 7 - 44983942 - 44986638 [-] on Build GRCh38
          GenBankmRNA BF317113.1 - 2, 3 215 Chr. 7 - 44983055 - 44986653 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0