Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG15 SIRNA Assays ›
          Gene Symbol:
          SNHG15
          Gene Name:
          small nucleolar RNA host gene 15
          Gene Aliases:
          C7orf40, Linc-Myo1g, MYO1GUT
          Chromosome Location:
          Chr.7: 44983028 - 44986660 on Build GRCh38
          UniGene ID:
          Hs.25892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270575

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003697.1 5 704
          GenBank mRNA AK096179.1 - 2 3012
          BC092459.1 - 5 704
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB560851.1 - 2 349 Chr. 7 - 44983026 - 44983974 [-] on Build GRCh38
          GenBankmRNA BF306650.1 - 633 Chr. - - [-] on Build
          GenBankmRNA CF127244.1 - 348 Chr. - - [-] on Build
          GenBankmRNA BF204739.1 - 5 703 Chr. 7 - 44983092 - 44986658 [-] on Build GRCh38
          GenBankmRNA BM825153.1 - 2 479 Chr. 7 - 44983026 - 44984104 [-] on Build GRCh38
          GenBankmRNA BG491747.1 - 5 726 Chr. 7 - 44983061 - 44986681 [-] on Build GRCh38
          GenBankmRNA HX788853.1 - 5 693 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA AB371433.1 - 5 682 Chr. 7 - 44983029 - 44986637 [-] on Build GRCh38
          GenBankmRNA HX785737.1 - 5 693 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BG748946.1 - 5 669 Chr. 7 - 44983065 - 44986624 [-] on Build GRCh38
          GenBankmRNA BE888491.1 - 648 Chr. - - [-] on Build
          GenBankmRNA BU502253.1 - 5 651 Chr. 7 - 44983028 - 44986604 [-] on Build GRCh38
          GenBankmRNA BQ434731.1 - 5 683 Chr. 7 - 44983024 - 44986638 [-] on Build GRCh38
          GenBankmRNA DB553836.1 - 2 334 Chr. 7 - 44983026 - 44983961 [-] on Build GRCh38
          GenBankmRNA DB563548.1 - 2 346 Chr. 7 - 44983026 - 44983970 [-] on Build GRCh38
          GenBankmRNA BI907654.1 - 4 630 Chr. 7 - 44983026 - 44986656 [-] on Build GRCh38
          GenBankmRNA BG026375.1 - 490 Chr. - - [-] on Build
          GenBankmRNA CD523041.1 - 2 272 Chr. 7 - 44983019 - 44983896 [-] on Build GRCh38
          GenBankmRNA CN265181.1 - 2 450 Chr. 7 - 44983033 - 44984039 [-] on Build GRCh38
          GenBankmRNA AA488149.1 - 3 466 Chr. 7 - 44983085 - 44986570 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4lrsz:80/100.66.79.169:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0