Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC286359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026847.1 | 4 | 473 | |||
GenBank mRNA | AK310296.1 | - | 4 | 473 | ||
AL833510.1 | - | 3 | 1535 |