Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLEU1 SIRNA Assays ›
          Gene Symbol:
          DLEU1
          Gene Name:
          deleted in lymphocytic leukemia 1 (non-protein coding)
          Gene Aliases:
          BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6
          Chromosome Location:
          Chr.13: 50082169 - 50528643 on Build GRCh38
          UniGene ID:
          Hs.591229
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272245

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLEU1
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002605.2
          1 262
          NR_109973.1
          1 262
          NR_109974.1
          1 262
          GenBank mRNA AJ412023.1 - LIT1929
          BCMS
          1 260
          AJ412024.1 - LIT1928
          BCMS
          1 260
          AJ412025.1 - LIT1927
          BCMS
          1 260
          AJ412026.1 - LIT1926
          BCMS
          1 260
          AJ412038.1 - LIT1907
          BCMS
          1 260
          AJ412039.1 - LIT1906
          BCMS
          1 260
          AJ412054.1 - LIT1886
          BCMS
          1 260
          AJ412055.1 - LIT1885
          BCMS
          1 260
          AJ412056.1 - LIT1884
          BCMS
          1 260
          AJ412057.1 - LIT1876
          BCMS
          1 260
          AJ412058.1 - LIT1875
          BCMS
          1 260
          AJ412059.1 - LIT1874
          BCMS
          1 260
          AJ412060.1 - LIT1873
          BCMS
          1 260
          AJ412061.1 - LIT1872
          BCMS
          1 260
          AJ412062.1 - LIT1871
          BCMS
          1 260
          AK095311.1 -
          1 182
          DB444728.1 -
          1 260
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA W05790.1 - 1 155 Chr. 13 - 50082276 - 50105047 [+] on Build GRCh38
          GenBankmRNA AF279660.3 - 5 85374 Chr. 13 - 49996975 - 50344526 [+] on Build GRCh38
          GenBankmRNA AJ411973.3 - 1 260 Chr. 13 - 50082171 - 50082556 [+] on Build GRCh38
          GenBankmRNA BM462952.1 - 233 Chr. - - [-] on Build
          GenBankmRNA DN991866.1 - 1 250 Chr. 13 - 50082188 - 50104959 [+] on Build GRCh38
          GenBankmRNA DB149054.1 - 222 Chr. - - [-] on Build
          GenBankmRNA CR741060.1 - 1 110 Chr. 13 - 50082321 - 50104891 [+] on Build GRCh38
          GenBankmRNA HY018670.1 - 1 242 Chr. 13 - 50082190 - 50104785 [+] on Build GRCh38
          GenBankmRNA BG527598.1 - 1 208 Chr. 13 - 50082223 - 50105056 [+] on Build GRCh38
          GenBankmRNA BQ212948.1 - 178 Chr. - - [-] on Build
          GenBankmRNA CR738820.1 - 1 110 Chr. 13 - 50082321 - 50104876 [+] on Build GRCh38
          GenBankmRNA BU787701.1 - 1 256 Chr. 13 - 50082178 - 50104929 [+] on Build GRCh38
          GenBankmRNA DA442398.1 - 1 182 Chr. 13 - 50082249 - 50104932 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0