Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO15B SIRNA Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          UniGene ID:
          Hs.390817
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n296590

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001309242.1 NP_001296171.1 24, 25 4519
          XM_017025120.1 XP_016880609.1 26, 27 4226
          XM_017025121.1 XP_016880610.1 26, 27 4211
          XM_017025122.1 XP_016880611.1 26, 27 4190
          XM_017025123.1 XP_016880612.1 26, 27 4228
          XM_017025124.1 XP_016880613.1 25, 26 4100
          XM_017025125.1 XP_016880614.1 24, 25 3949
          XM_017025126.1 XP_016880615.1 21, 22 3691
          XM_017025127.1 XP_016880616.1 24, 25 3534
          XM_017025128.1 XP_016880617.1 25, 26 3280
          XM_017025129.1 XP_016880618.1 24, 25 2512
          XM_017025130.1 XP_016880619.1 23, 24 2468
          XM_017025131.1 XP_016880620.1 22, 23 2907
          XM_017025132.1 XP_016880621.1 21, 22 2848
          XM_017025133.1 XP_016880622.1 19, 20 2373
          XM_017025134.1 XP_016880623.1 20, 21 2734
          XM_017025135.1 XP_016880624.1 14, 15 1390
          XM_017025136.1 XP_016880625.1 14, 15 1739
          XM_017025137.1 XP_016880626.1 11, 12 1284
          XM_017025138.1 XP_016880627.1 6, 7 780
          XM_017025139.1 XP_016880628.1 7, 8 581
          XM_017025140.1 XP_016880629.1 4, 5 394
          XM_017025141.1 XP_016880630.1 26, 27 4492
          XM_017025142.1 XP_016880631.1 26, 27 4492
          XM_017025144.1 XP_016880633.1 19, 20 4101
          GenBank mRNA AB058686.1 - 14, 15 2626
          AF418286.1 - 22, 23 4752
          AF418289.1 - 19, 20 4101
          AF418290.1 - 19, 20 4947
          AF418291.1 - 20, 21 4102
          AK096869.1 - 3, 4 227
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE767401.1 - 4, 5 351 Chr. 17 - 75610193 - 75612864 [+] on Build GRCh38
          GenBankmRNA HY166912.1 - 4, 5 303 Chr. 17 - 75610676 - 75613077 [+] on Build GRCh38
          GenBankmRNA BM906425.1 - 327 Chr. - - [-] on Build
          GenBankmRNA BM738080.1 - 3, 4 239 Chr. 17 - 75610915 - 75613211 [+] on Build GRCh38
          GenBankmRNA BM737684.1 - 4, 5 272 Chr. 17 - 75610707 - 75613190 [+] on Build GRCh38
          GenBankmRNA BX401175.2 - 3, 4 248 Chr. 17 - 75610899 - 75614270 [+] on Build GRCh38
          GenBankmRNA DA890157.1 - 3, 4 227 Chr. 17 - 75610922 - 75613211 [+] on Build GRCh38
          GenBankmRNA BM738296.1 - 4, 5 280 Chr. 17 - 75610699 - 75613107 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0