Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306215.1 | NP_001293144.1 | 15 | 5924 | ||
NM_178835.4 | NP_849157.2 | 15 | 5933 | |||
XM_011531634.2 | XP_011529936.1 | 14 | 5938 | |||
XM_011531635.2 | XP_011529937.1 | 15 | 11425 | |||
XM_011531645.2 | XP_011529947.1 | 10 | 3676 | |||
XM_017007768.1 | XP_016863257.1 | 14 | 5933 | |||
XM_017007769.1 | XP_016863258.1 | 14 | 12184 | |||
XM_017007770.1 | XP_016863259.1 | 13 | 5705 | |||
XM_017007777.1 | XP_016863266.1 | 11 | 3675 | |||
GenBank mRNA | AL117519.1 | - | 1 | 631 | ||
BC033248.1 | - | 1 | 1087 |