Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138646.3 | NP_619587.3 | 12 | 2232 | ||
GenBank mRNA | AK081489.1 | - | 11 | 2586 | ||
AK129418.1 | - | 11 | 2119 | |||
AK147954.1 | - | 11 | 2675 | |||
AK171871.1 | - | 12 | 2201 | |||
AK185532.1 | - | 2 | 262 | |||
AY043415.1 | - | 11 | 1752 | |||
BC004668.1 | - | 2 | 839 | |||
BC063055.1 | - | 11 | 1924 | 5701206 |