Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199632.1 | NP_001186561.1 | 17 | 1893 | ||
NM_177076.4 | NP_796050.2 | 16 | 1794 | |||
GenBank mRNA | AK029566.1 | - | 16 | 1796 | ||
AK030172.1 | - | 17 | 1877 | |||
BC145291.1 | - | 17 | 1819 | 9053845 | ||
BC145292.1 | - | 16 | 1721 | 9053846 |