Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PVALB SIRNA Assays ›
Gene Symbol:
Pvalb
Gene Name:
parvalbumin
Gene Aliases:
PALB1, Pva
Chromosome Location:
Chr.7: 119428652 - 119443656 on Build Rnor_6.0
UniGene ID:
Rn.2005
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s129329
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pvalb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022499.2
NP_071944.1
2
136
XM_006241929.2
XP_006241991.1
3
209
GenBank mRNA
BC126090.1
-
2
136
7321065
FQ212120.1
-
3
199
FQ214597.1
-
3
197
FQ214667.1
-
3
198
FQ214909.1
-
3
197
FQ215072.1
-
3
197
FQ215101.1
-
1
33
FQ215172.1
-
3
197
FQ215177.1
-
3
196
FQ215250.1
-
3
198
FQ215353.1
-
3
197
FQ215367.1
-
3
197
FQ215388.1
-
3
197
FQ215455.1
-
3
197
FQ215472.1
-
3
197
FQ215487.1
-
3
197
FQ215608.1
-
3
198
FQ215681.1
-
3
197
FQ216147.1
-
3
197
FQ216186.1
-
3
197
FQ216201.1
-
3
198
FQ216260.1
-
3
197
FQ216338.1
-
3
198
FQ216339.1
-
3
197
FQ216448.1
-
3
197
FQ216459.1
-
3
197
FQ216533.1
-
3
197
FQ216789.1
-
3
197
FQ216830.1
-
3
197
FQ216843.1
-
3
197
FQ216867.1
-
3
197
FQ216872.1
-
3
197
FQ216903.1
-
3
199
FQ216906.1
-
3
194
FQ216960.1
-
3
197
FQ216981.1
-
3
197
FQ217053.1
-
3
197
FQ217064.1
-
3
197
FQ217099.1
-
3
197
FQ217132.1
-
2
198
FQ217209.1
-
3
196
FQ217210.1
-
3
197
FQ217278.1
-
3
197
FQ217298.1
-
3
198
FQ217358.1
-
3
197
FQ217373.1
-
3
198
FQ217384.1
-
3
197
FQ217418.1
-
3
198
FQ217424.1
-
3
197
FQ217446.1
-
2
165
FQ217489.1
-
3
197
FQ217527.1
-
3
197
FQ217562.1
-
3
197
FQ217576.1
-
3
197
FQ217645.1
-
3
196
FQ217652.1
-
3
198
FQ217677.1
-
3
198
FQ217743.1
-
3
196
FQ217749.1
-
3
198
FQ217760.1
-
3
198
FQ217770.1
-
3
197
FQ217787.1
-
3
198
FQ217845.1
-
3
197
FQ217858.1
-
3
197
FQ217860.1
-
3
198
FQ217861.1
-
3
197
FQ217880.1
-
3
199
FQ217886.1
-
3
197
FQ217934.1
-
2
139
FQ217966.1
-
3
197
FQ217976.1
-
3
197
FQ217991.1
-
3
197
FQ218035.1
-
3
197
FQ218057.1
-
3
197
FQ223582.1
-
3
197
FQ223596.1
-
3
197
FQ223603.1
-
3
198
FQ223630.1
-
3
194
FQ223664.1
-
3
198
FQ223669.1
-
1
31
FQ223695.1
-
3
197
FQ223712.1
-
3
198
FQ223861.1
-
3
197
FQ223863.1
-
3
197
FQ223941.1
-
3
197
FQ223961.1
-
3
197
FQ223988.1
-
3
196
FQ223989.1
-
3
197
FQ223990.1
-
3
197
FQ224014.1
-
3
197
FQ224025.1
-
3
198
FQ224028.1
-
3
197
FQ224037.1
-
3
197
FQ224053.1
-
3
197
FQ224059.1
-
3
197
FQ224083.1
-
3
197
FQ224113.1
-
3
197
FQ224136.1
-
3
197
FQ224173.1
-
3
197
FQ224181.1
-
3
197
FQ224187.1
-
3
197
FQ224242.1
-
3
198
FQ224248.1
-
3
195
FQ224294.1
-
3
197
FQ224303.1
-
3
198
FQ224322.1
-
3
197
FQ224356.1
-
3
197
FQ224363.1
-
3
197
FQ224417.1
-
3
197
FQ224430.1
-
3
197
FQ224435.1
-
3
195
FQ224463.1
-
3
198
FQ224483.1
-
3
197
FQ224485.1
-
3
197
FQ224488.1
-
3
198
FQ224510.1
-
3
197
FQ224541.1
-
3
197
FQ224580.1
-
3
197
FQ224584.1
-
3
198
FQ224600.1
-
3
197
FQ224613.1
-
3
197
FQ224634.1
-
3
197
FQ224643.1
-
3
197
FQ224673.1
-
3
198
FQ224723.1
-
3
199
FQ224767.1
-
3
197
FQ224773.1
-
3
199
FQ224778.1
-
3
197
FQ224913.1
-
3
197
M10764.1
-
1
22
M12725.1
-
3
196
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_002854, BC096112, BC096113, BC096115, AK311832, and others.
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytosolic calcium ion concentration
calcium ion binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5688f:80/100.66.73.65:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0