Search
Search

F10| Gene Symbol: F10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017143.2 | NP_058839.1 | 4 | 450 | ||
| GenBank mRNA | BC088151.1 | - | 4 | 450 | 7383301 | |
| FQ211285.1 | - | 4 | 374 | |||
| FQ219425.1 | - | 4 | 368 | |||
| X79807.1 | - | 4 | 362 | |||