Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FTL1 SIRNA Assays ›
          Gene Symbol:
          Ftl1 LOC100360087
          Gene Name:
          ferritin light chain 1
          ferritin light chain 1-like
          Gene Aliases:
          Ftl
          -
          Chromosome Location:
          Chr.1: 101448190 - 101450034 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 152325856 - 152326797 on Build Rnor_6.0
          UniGene ID:
          Rn.1905
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s131112

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ftl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022500.4 NP_071945.3 2 411
          GenBank mRNA BC061525.1 - 2 415 6921603
          BC086583.1 - 2 392 7307718
          BC088756.1 - 2 392 7307747
          FQ209694.1 - 2 411
          FQ209816.1 - 2 411
          FQ218577.1 - 2 411
          FQ219483.1 - 2 411
          FQ219685.1 - 2 411
          FQ219809.1 - 2 411
          FQ219930.1 - 2 410
          FQ220095.1 - 2 411
          FQ220123.1 - 2 411
          FQ220243.1 - 2 408
          FQ220651.1 - 2 411
          FQ220814.1 - 2 411
          FQ220884.1 - 2 411
          FQ221898.1 - 2 411
          FQ222411.1 - 1 315
          FQ223227.1 - 2 411
          FQ226019.1 - 2 411
          FQ226254.1 - 2 367
          FQ226458.1 - 2 410
          FQ226986.1 - 2 407
          FQ227401.1 - 2 411
          FQ227800.1 - 2 411
          FQ227843.1 - 2 411
          FQ228050.1 - 2 411
          FQ228585.1 - 2 411
          FQ229176.1 - 2 411
          FQ229284.1 - 2 411
          FQ229370.1 - 2 411
          FQ229673.1 - 2 409
          FQ229757.1 - 2 411
          FQ230098.1 - 2 409
          FQ230445.1 - 2 411
          FQ230659.1 - 2 407
          FQ231322.1 - 2 411
          FQ231373.1 - 2 411
          FQ231415.1 - 1 267
          FQ231464.1 - 2 411
          FQ231570.1 - 2 411
          FQ231615.1 - 2 411
          FQ231781.1 - 2 407
          FQ231957.1 - 2 411
          FQ232031.1 - 2 411
          FQ232167.1 - 2 411
          FQ232317.1 - 2 367
          FQ232517.1 - 2 411
          FQ233772.1 - 2 411
          FQ234074.1 - 2 411
          FQ234603.1 - 2 411
          FQ234647.1 - 2 411
          FQ235044.1 - 2 411
          FQ235250.1 - 2 411
          K01930.1 - 2 336
          L01122.1 - 2 364
          Gene Symbol: LOC100360087
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002729553.4 XP_002729599.1 1 426

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: S89400.

          Panther Classification:

          Molecular Function -

          storage protein

          Gene Ontology Categories:

          Function(s) Process(es)

          iron ion transport
          cellular iron ion homeostasis
          response to lead ion
          molecular_function
          iron ion binding
          ferric iron binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5ps9:80/100.66.76.38:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0