Search
Search

Cdk1| Gene Symbol: Cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019296.1 | NP_062169.1 | 2, 3 | 146 | ||
| XM_006256353.2 | XP_006256415.1 | 3, 4 | 273 | |||
| GenBank mRNA | BC091549.1 | - | 3, 4 | 231 | 7388690 | |
| FQ219810.1 | - | 2, 3 | 149 | |||
| FQ227059.1 | - | 2, 3 | 149 | |||
| X60767.1 | - | 2, 3 | 146 | |||