Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRF SIRNA Assays ›
          Gene Symbol:
          SRF
          Gene Name:
          serum response factor
          Gene Aliases:
          MCM1
          Chromosome Location:
          Chr.6: 43171295 - 43181506 on Build GRCh38
          UniGene ID:
          Hs.520140
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13427

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001292001.1 NP_001278930.1 5 777
          NM_003131.3 NP_003122.1 5 1590
          GenBank mRNA AB209128.1 - 3 4565
          AK300467.1 - 5 777
          BC048211.1 - 5 1426 5808348
          BC052572.1 - 5 1590 6462845
          J03161.1 - 5 1586

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          patterning of blood vessels
          response to hypoxia
          mesoderm formation
          neuron migration
          trophectodermal cell differentiation
          heart looping
          morphogenesis of an epithelial sheet
          cell migration involved in sprouting angiogenesis
          positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
          transcription from RNA polymerase II promoter
          cell-matrix adhesion
          heart development
          long-term memory
          negative regulation of cell proliferation
          associative learning
          response to toxic substance
          response to hormone
          epithelial structure maintenance
          positive regulation of transcription via serum response element binding
          hippocampus development
          tangential migration from the subventricular zone to the olfactory bulb
          regulation of cell adhesion
          platelet activation
          platelet formation
          negative regulation of cell migration
          thyroid gland development
          neuron projection development
          regulation of water loss via skin
          response to cytokine
          megakaryocyte development
          dorsal aorta morphogenesis
          mRNA transcription from RNA polymerase II promoter
          stress fiber assembly
          skin morphogenesis
          positive thymic T cell selection
          sarcomere organization
          positive regulation of cell differentiation
          positive regulation of axon extension
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle contraction
          positive regulation of transcription by glucose
          muscle cell cellular homeostasis
          thymus development
          developmental growth
          neuron development
          erythrocyte development
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of smooth muscle cell differentiation
          positive regulation of filopodium assembly
          cardiac myofibril assembly
          angiogenesis involved in wound healing
          hematopoietic stem cell differentiation
          positive regulation of transcription initiation from RNA polymerase II promoter
          long term synaptic depression
          face development
          heart trabecula formation
          lung morphogenesis
          bronchus cartilage development
          trachea cartilage development
          cardiac vascular smooth muscle cell differentiation
          eyelid development in camera-type eye
          lung smooth muscle development
          bicellular tight junction assembly
          cellular response to glucose stimulus
          primitive streak formation
          epithelial cell-cell adhesion
          cellular senescence
          negative regulation of beta-amyloid clearance
          negative regulation of pri-miRNA transcription from RNA polymerase II promoter
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          transcription factor activity, RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          transcription factor binding
          serum response element binding
          chromatin DNA binding
          protein homodimerization activity
          histone deacetylase binding
          primary miRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7l4vz:80/100.66.76.145:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0