Search
Search

| Gene Symbol: Lenep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053614.1 | NP_446066.1 | 1 | 154 | ||
| GenBank mRNA | U15149.1 | - | 1 | 154 | ||
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110138.1 | NP_001103608.1 | 8 | 2324 | ||