Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004199.1 | NP_001004199.1 | 11 | 1510 | ||
XM_006236480.2 | XP_006236542.1 | 11 | 1553 | |||
XM_006236481.2 | XP_006236543.1 | 11 | 1996 | |||
XM_006236482.2 | XP_006236544.1 | 11 | 1553 | |||
XM_006236483.2 | XP_006236545.1 | 11 | 1553 | |||
GenBank mRNA | AF509819.1 | - | 5 | 686 | ||
AY318959.1 | - | 1426 | ||||
BC081949.1 | - | 11 | 1510 | 7122283 |