Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033895.1 | NP_001029067.1 | 12 | 1429 | ||
XM_008761146.1 | XP_008759368.1 | 11 | 1474 | |||
GenBank mRNA | BC128708.1 | - | 12 | 1554 | ||
BC168242.1 | - | 12 | 1542 | 7934746 | ||
DQ185515.1 | - | 12 | 1429 |