Search
Search

Ak8| Gene Symbol: Ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004266.1 | NP_001004266.1 | 12, 13 | 1219 | ||
| XM_006233794.2 | XP_006233856.1 | 13, 14 | 1255 | |||
| GenBank mRNA | BC079446.1 | - | 12, 13 | 1219 | 7115585 | |