Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Smug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177934.3 | NP_808795.1 | 3 | 557 | ||
XM_006242399.2 | XP_006242461.1 | 2 | 378 | |||
XM_008765749.1 | XP_008763971.1 | 3 | 434 | |||
GenBank mRNA | AY191521.1 | - | 3 | 562 |